Assay Details
| Gene Symbol: WRNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554172.2 | XP_005554229.1 | 5 - 6 | 1882 | 140 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

WRNIP1| Gene Symbol: WRNIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554172.2 | XP_005554229.1 | 5 - 6 | 1882 | 140 | |
Set Membership: |
|