Assay Details
| Gene Symbol: GPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577349.2 | XP_005577406.1 | 2 - 3 | 599 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GPHA2| Gene Symbol: GPHA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577349.2 | XP_005577406.1 | 2 - 3 | 599 | 64 | |
Set Membership: |
|