Assay Details
| Gene Symbol: SHROOM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005557750.2 | XP_005557807.1 | 2 - 3 | 2105 | 143 | |
| XM_005557752.2 | XP_005557809.1 | 2 - 3 | 1999 | 143 | ||
More Information
Set Membership: |
|

SHROOM1
Genomic Map