Assay Details
| Gene Symbol: SHF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005559429.2 | XP_005559486.1 | 6 - 7 | 1100 | 106 | |
| XM_005559430.2 | XP_005559487.1 | 4 - 5 | 739 | 106 | ||
More Information
Set Membership: |
|

SHF
Genomic Map