Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551918.2 | XP_005551975.1 | 5 - 6 | 4165 | 101 | |
| XM_005551919.2 | XP_005551976.1 | 5 - 6 | 3878 | 101 | ||
| XM_005551920.2 | XP_005551977.1 | 4 - 5 | 490 | 101 | ||
| XM_015448709.1 | XP_015304195.1 | 4 - 5 | 568 | 101 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map