Assay Details
| Gene Symbol: MLIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552707.2 | XP_005552764.1 | 12 - 13 | 4527 | 130 | |
| XM_005552708.2 | XP_005552765.2 | 12 - 13 | 3030 | 130 | ||
| XM_005552709.2 | XP_005552766.2 | 12 - 13 | 3596 | 130 | ||
| XM_005552713.2 | XP_005552770.2 | 11 - 12 | 2876 | 130 | ||
| XM_005552717.2 | XP_005552774.2 | 11 - 12 | 1443 | 130 | ||
| XM_015449298.1 | XP_015304784.1 | 12 - 13 | 3104 | 130 | ||
| XM_015449299.1 | XP_015304785.1 | 11 - 12 | 4446 | 130 | ||
| XM_015449300.1 | XP_015304786.1 | 10 - 11 | 4386 | 130 | ||
| XM_015449301.1 | XP_015304787.1 | 11 - 12 | 4374 | 130 | ||
| XM_015449303.1 | XP_015304789.1 | 9 - 10 | 4324 | 130 | ||
| XM_015449304.1 | XP_015304790.1 | 8 - 9 | 4280 | 130 | ||
| XM_015449305.1 | XP_015304791.1 | 13 - 14 | 3010 | 130 | ||
| XM_015449306.1 | XP_015304792.1 | 13 - 14 | 3006 | 130 | ||
More Information
Set Membership: |
|

MLIP
Genomic Map