Assay Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005553011.2 | XP_005553068.1 | 6 - 7 | 657 | 147 | |
| XM_005553012.2 | XP_005553069.1 | 6 - 7 | 657 | 147 | ||
| XM_005553013.2 | XP_005553070.1 | 6 - 7 | 657 | 147 | ||
More Information
Set Membership: |
|

TAF8
Genomic Map