Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540876.2 | XP_005540933.1 | 4 - 5 | 519 | 127 | |
| XM_005540877.2 | XP_005540934.1 | 4 - 5 | 843 | 127 | ||
| XM_005540878.2 | XP_005540935.1 | 4 - 5 | 617 | 127 | ||
| GenBank mRNA | AB171582.1 | - | 3 - 4 | 328 | 127 | |
More Information
Set Membership: |
|

LYPLAL1
Genomic Map