Assay Details
Gene Symbol: SLC9B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319594.1 | NP_001306523.1 | 13 - 14 | 1476 | 122 | |
XM_005555563.2 | XP_005555620.1 | 9 - 10 | 1196 | 122 | ||
XM_005555564.2 | XP_005555621.1 | 9 - 10 | 1213 | 122 | ||
XM_015450707.1 | XP_015306193.1 | 11 - 12 | 1857 | 122 | ||
XM_015450708.1 | XP_015306194.1 | 12 - 13 | 1393 | 122 | ||
XM_015450709.1 | XP_015306195.1 | 10 - 11 | 1220 | 122 | ||
XM_015450710.1 | XP_015306196.1 | 10 - 11 | 1281 | 122 | ||
XM_015450711.1 | XP_015306197.1 | 13 - 14 | 1520 | 122 | ||
GenBank mRNA | AB070105.1 | - | 12 - 13 | 1336 | 122 | |
AB168740.1 | - | 13 - 14 | 1476 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
