Assay Details
| Gene Symbol: SLC39A11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584820.2 | XP_005584877.1 | 6 - 7 | 738 | 88 | |
| XM_005584821.2 | XP_005584878.1 | 6 - 7 | 717 | 88 | ||
| XM_015438786.1 | XP_015294272.1 | 7 - 8 | 1106 | 88 | ||
| XM_015438787.1 | XP_015294273.1 | 6 - 7 | 995 | 88 | ||
| XM_015438788.1 | XP_015294274.1 | 6 - 7 | 738 | 88 | ||
More Information
Set Membership: |
|

SLC39A11
Genomic Map