Assay Details
| Gene Symbol: ORMDL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005584053.2 | XP_005584110.1 | 2 - 3 | 470 | 118 | |
| XM_005584054.2 | XP_005584111.1 | 2 - 3 | 505 | 118 | ||
| XM_015438438.1 | XP_015293924.1 | 2 - 3 | 1007 | 118 | ||
| XM_015438439.1 | XP_015293925.1 | 2 - 3 | 355 | 118 | ||
| XM_015438440.1 | XP_015293926.1 | 2 - 3 | 220 | 118 | ||
More Information
Set Membership: |
|

ORMDL3
Genomic Map