Assay Details
| Gene Symbol: RHEBL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570748.2 | XP_005570805.1 | 3 - 4 | 418 | 120 | |
| XM_005570750.2 | XP_005570807.1 | 3 - 4 | 486 | 120 | ||
| XM_015430873.1 | XP_015286359.1 | 3 - 4 | 346 | 120 | ||
| XM_015430874.1 | XP_015286360.1 | 4 - 5 | 331 | 120 | ||
More Information
Set Membership: |
|

RHEBL1
Genomic Map