Assay Details
| Gene Symbol: CAPSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556717.2 | XP_005556774.1 | 2 - 3 | 302 | 114 | |
| XM_005556718.2 | XP_005556775.1 | 2 - 3 | 224 | 114 | ||
| XM_015451272.1 | XP_015306758.1 | 2 - 3 | 244 | 114 | ||
More Information
Set Membership: |
|

CAPSL
Genomic Map