Assay Details
| Gene Symbol: ROR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582234.2 | XP_005582291.1 | 1 - 2 | 408 | 95 | |
| XM_015436575.1 | XP_015292061.1 | 1 - 2 | 408 | 95 | ||
More Information
Set Membership: |
|

ROR2
Genomic Map