Assay Details
| Gene Symbol: OVOL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005577237.2 | XP_005577294.1 | 2 - 3 | 732 | 114 | |
| XM_005577238.2 | XP_005577295.1 | 2 - 3 | 667 | 114 | ||
| XM_015434449.1 | XP_015289935.1 | 2 - 3 | 2029 | 114 | ||
More Information
Set Membership: |
|

OVOL1
Genomic Map