Assay Details
| Gene Symbol: BAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005547336.2 | XP_005547393.1 | 8 - 9 | 885 | 80 | |
| XM_005547337.2 | XP_005547394.1 | 8 - 9 | 885 | 80 | ||
| XM_005547338.2 | XP_005547395.1 | 8 - 9 | 885 | 80 | ||
More Information
Set Membership: |
|

BAP1
Genomic Map