Assay Details
| Gene Symbol: ASIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551179.2 | XP_005551236.1 | 11 - 12 | 4021 | 158 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASIC3| Gene Symbol: ASIC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005551179.2 | XP_005551236.1 | 11 - 12 | 4021 | 158 | |
Set Membership: |
|