Assay Details
| Gene Symbol: PPIG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005573416.2 | XP_005573473.1 | 7 - 8 | 595 | 144 | |
| XM_005573417.2 | XP_005573474.1 | 7 - 8 | 795 | 144 | ||
More Information
Set Membership: |
|

PPIG
Genomic Map