Assay Details
| Gene Symbol: MECP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284547.1 | NP_001271476.1 | 2 - 3 | 382 | 89 | |
| GenBank mRNA | AF295597.1 | - | 2 - 3 | 382 | 89 | |
More Information
Set Membership: |
|

MECP2
Genomic Map