Assay Details
| Gene Symbol: ROR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015434778.1 | XP_015290264.1 | 7 - 8 | 2229 | 143 | |
| XM_015434786.1 | XP_015290272.1 | 8 - 9 | 5338 | 143 | ||
| XM_015434792.1 | XP_015290278.1 | 9 - 10 | 2910 | 143 | ||
| XM_015434797.1 | XP_015290283.1 | 7 - 8 | 5242 | 143 | ||
| XM_015434801.1 | XP_015290287.1 | 9 - 10 | 4272 | 143 | ||
| XM_015434806.1 | XP_015290292.1 | 8 - 9 | 4176 | 143 | ||
| XM_015434816.1 | XP_015290302.1 | 7 - 8 | 4012 | 143 | ||
More Information
Set Membership: |
|

ROR1
Genomic Map