Assay Details
| Gene Symbol: PIP4K2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005564792.2 | XP_005564849.1 | 5 - 6 | 775 | 83 | |
| XM_005564793.2 | XP_005564850.1 | 4 - 5 | 622 | 83 | ||
| XM_015455645.1 | XP_015311131.1 | 5 - 6 | 1106 | 83 | ||
More Information
Set Membership: |
|

PIP4K2A
Genomic Map