Assay Details
| Gene Symbol: CBFA2T3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015443170.1 | XP_015298656.1 | 4 - 5 | 3822 | 94 | |
| XM_015443171.1 | XP_015298657.1 | 3 - 4 | 3747 | 94 | ||
More Information
Set Membership: |
|

CBFA2T3
Genomic Map