Assay Details
| Gene Symbol: FGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544263.2 | XP_005544320.1 | 6 - 7 | 763 | 87 | |
| XM_005544264.2 | XP_005544321.1 | 6 - 7 | 1093 | 87 | ||
| XM_005544265.2 | XP_005544322.1 | 6 - 7 | 634 | 87 | ||
| XM_015439923.1 | XP_015295409.1 | 5 - 6 | 664 | 87 | ||
More Information
Set Membership: |
|

FGR
Genomic Map