Assay Details
| Gene Symbol: FCMR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005540654.2 | XP_005540711.1 | 5 - 6 | 1102 | 168 | |
| XM_015449203.1 | XP_015304689.1 | 5 - 6 | 1102 | 168 | ||
| XM_015449213.1 | XP_015304699.1 | 4 - 5 | 572 | 168 | ||
More Information
Set Membership: |
|

FCMR
Genomic Map