Assay Details
| Gene Symbol: WDR46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283915.1 | NP_001270844.1 | 10 - 11 | 1150 | 72 | |
| XM_015448786.1 | XP_015304272.1 | 10 - 11 | 1190 | 72 | ||
| GenBank mRNA | AB168376.1 | - | 10 - 11 | 1141 | 72 | |
| AB173545.1 | - | 10 - 11 | 1150 | 72 | ||
More Information
Set Membership: |
|

WDR46
Genomic Map