Assay Details
| Gene Symbol: BCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005570373.2 | XP_005570430.2 | 3 - 4 | 410 | 72 | |
| XM_005570374.2 | XP_005570431.1 | 3 - 4 | 817 | 72 | ||
| XM_005570375.2 | XP_005570432.1 | 3 - 4 | 753 | 72 | ||
| XM_015430502.1 | XP_015285988.1 | 3 - 4 | 411 | 72 | ||
More Information
Set Membership: |
|

BCAT1
Genomic Map