Assay Details
| Gene Symbol: LGALS3BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585146.2 | XP_005585203.1 | 5 - 6 | 934 | 151 | |
| XM_005585147.2 | XP_005585204.1 | 5 - 6 | 936 | 151 | ||
| XM_015438918.1 | XP_015294404.1 | 5 - 6 | 959 | 151 | ||
More Information
Set Membership: |
|

LGALS3BP
Genomic Map