Assay Details
| Gene Symbol: SIGMAR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581448.2 | XP_005581505.1 | 3 - 4 | 1134 | 84 | |
| XM_015437058.1 | XP_015292544.1 | 2 - 3 | 136 | 84 | ||
More Information
Set Membership: |
|

SIGMAR1
Genomic Map