Assay Details
| Gene Symbol: LOC102142055 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588526.2 | XP_005588583.1 | 3 - 4 | 1525 | 94 | |
| XM_005588527.2 | XP_005588584.1 | 3 - 4 | 1585 | 94 | ||
| XM_005588529.2 | XP_005588586.1 | 3 - 4 | 458 | 94 | ||
| XM_015440712.1 | XP_015296198.1 | 6 - 7 | 700 | 94 | ||
| XM_015440713.1 | XP_015296199.1 | 7 - 8 | 767 | 94 | ||
| XM_015440714.1 | XP_015296200.1 | 5 - 6 | 588 | 94 | ||
| XM_015440715.1 | XP_015296201.1 | 5 - 6 | 589 | 94 | ||
More Information
Set Membership: |
|

LOC102142055
Genomic Map