Assay Details
| Gene Symbol: PRKAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005572377.2 | XP_005572434.1 | 2 - 3 | 817 | 98 | |
| XM_005572378.2 | XP_005572435.1 | 2 - 3 | 817 | 98 | ||
More Information
Set Membership: |
|

PRKAB1
Genomic Map