Assay Details
| Gene Symbol: TESK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581376.2 | XP_005581433.1 | 5 - 6 | 937 | 130 | |
| XM_015437021.1 | XP_015292507.1 | 5 - 6 | 937 | 130 | ||
More Information
Set Membership: |
|

TESK1
Genomic Map