Assay Details
| Gene Symbol: LOC102143204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565127.2 | XP_005565184.1 | 3 - 4 | 438 | 79 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102143204| Gene Symbol: LOC102143204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565127.2 | XP_005565184.1 | 3 - 4 | 438 | 79 | |
Set Membership: |
|