Assay Details
| Gene Symbol: ERF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589394.2 | XP_005589451.1 | 3 - 4 | 550 | 112 | |
| XM_015442078.1 | XP_015297564.1 | 3 - 4 | 3137 | 112 | ||
| XM_015442079.1 | XP_015297565.1 | 3 - 4 | 415 | 112 | ||
| XM_015442080.1 | XP_015297566.1 | 3 - 4 | 469 | 112 | ||
| XM_015442081.1 | XP_015297567.1 | 3 - 4 | 431 | 112 | ||
More Information
Set Membership: |
|

ERF
Genomic Map