Assay Details
| Gene Symbol: WASF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585522.2 | XP_005585579.1 | 4 - 5 | 486 | 82 | |
| XM_015439083.1 | XP_015294569.1 | 4 - 5 | 486 | 82 | ||
| XM_015439084.1 | XP_015294570.1 | 5 - 6 | 538 | 82 | ||
| XM_015439086.1 | XP_015294572.1 | 3 - 4 | 334 | 82 | ||
| XM_015439087.1 | XP_015294573.1 | 3 - 4 | 481 | 82 | ||
| XM_015439088.1 | XP_015294574.1 | 3 - 4 | 303 | 82 | ||
More Information
Set Membership: |
|

WASF3
Genomic Map