Assay Details
| Gene Symbol: PIM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285125.1 | NP_001272054.1 | 2 - 3 | 356 | 69 | |
| GenBank mRNA | AB173654.1 | - | 2 - 3 | 356 | 69 | |
More Information
Set Membership: |
|

PIM2
Genomic Map