Assay Details
| Gene Symbol: RXRG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005539844.2 | XP_005539901.1 | 1 - 2 | 626 | 59 | |
| GenBank mRNA | AB171038.1 | - | 2 - 3 | 218 | 59 | |
More Information
Set Membership: |
|

RXRG
Genomic Map