Assay Details
| Gene Symbol: NR4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581200.2 | XP_005581257.1 | 7 - 8 | 1806 | 90 | |
| XM_015436932.1 | XP_015292418.1 | 7 - 8 | 2593 | 90 | ||
More Information
Set Membership: |
|

NR4A3
Genomic Map