Assay Details
| Gene Symbol: CLDN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005586101.2 | XP_005586158.1 | 2 - 3 | 463 | 134 | |
| XM_005586102.2 | XP_005586159.1 | 2 - 3 | 635 | 134 | ||
| XM_005586103.2 | XP_005586160.1 | 2 - 3 | 578 | 134 | ||
More Information
Set Membership: |
|

CLDN10
Genomic Map