Assay Details
| Gene Symbol: PEMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005583028.2 | XP_005583085.2 | 4 - 5 | 390 | 133 | |
| XM_005583029.1 | XP_005583086.1 | 4 - 5 | 607 | 133 | ||
| XM_005583030.2 | XP_005583087.1 | 5 - 6 | 663 | 133 | ||
| XM_005583031.2 | XP_005583088.1 | 4 - 5 | 535 | 133 | ||
| XM_015437715.1 | XP_015293201.1 | 5 - 6 | 657 | 133 | ||
| XM_015437716.1 | XP_015293202.1 | 5 - 6 | 666 | 133 | ||
| XM_015437717.1 | XP_015293203.1 | 5 - 6 | 659 | 133 | ||
| XM_015437718.1 | XP_015293204.1 | 4 - 5 | 588 | 133 | ||
More Information
Set Membership: |
|

PEMT
Genomic Map