Assay Details
Gene Symbol: IRAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005571462.2 | XP_005571519.1 | 7 - 8 | 1187 | 123 | |
XM_005571463.2 | XP_005571520.1 | 6 - 7 | 711 | 123 | ||
XM_015431165.1 | XP_015286651.1 | 7 - 8 | 753 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
