Assay Details
| Gene Symbol: STAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555191.2 | XP_005555248.1 | 5 - 6 | 638 | 116 | |
| XM_005555192.2 | XP_005555249.1 | 5 - 6 | 641 | 116 | ||
More Information
Set Membership: |
|

STAP1
Genomic Map