Assay Details
| Gene Symbol: DDAH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542839.2 | XP_005542896.1 | 4 - 5 | 981 | 91 | |
| XM_005542843.2 | XP_005542900.1 | 4 - 5 | 577 | 91 | ||
| XM_015433415.1 | XP_015288901.1 | 5 - 6 | 1043 | 91 | ||
More Information
Set Membership: |
|

DDAH1
Genomic Map