Assay Details
| Gene Symbol: FBXL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552301.2 | XP_005552358.1 | 6 - 7 | 1610 | 92 | |
| XM_005552302.2 | XP_005552359.1 | 7 - 8 | 1633 | 92 | ||
| XM_005552303.2 | XP_005552360.1 | 5 - 6 | 1492 | 92 | ||
| XM_015449053.1 | XP_015304539.1 | 5 - 6 | 1777 | 92 | ||
More Information
Set Membership: |
|

FBXL4
Genomic Map