Assay Details
| Gene Symbol: SIRT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005554003.2 | XP_005554060.1 | 8 - 9 | 1039 | 121 | |
| XM_005554004.2 | XP_005554061.1 | 8 - 9 | 887 | 121 | ||
| XM_005554005.2 | XP_005554062.1 | 7 - 8 | 957 | 121 | ||
| XM_005554006.2 | XP_005554063.1 | 7 - 8 | 864 | 121 | ||
| XM_005554008.2 | XP_005554065.1 | 7 - 8 | 888 | 121 | ||
| XM_005554009.2 | XP_005554066.1 | 8 - 9 | 853 | 121 | ||
| XM_005554010.2 | XP_005554067.1 | 7 - 8 | 881 | 121 | ||
| XM_015449886.1 | XP_015305372.1 | 7 - 8 | 2507 | 121 | ||
| XM_015449889.1 | XP_015305375.1 | 6 - 7 | 2393 | 121 | ||
More Information
Set Membership: |
|

SIRT5
Genomic Map