Assay Details
| Gene Symbol: SLC35A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542613.2 | XP_005542670.1 | 7 - 8 | 1104 | 80 | |
| XM_005542614.2 | XP_005542671.1 | 7 - 8 | 2168 | 80 | ||
More Information
Set Membership: |
|

SLC35A3
Genomic Map