Assay Details
| Gene Symbol: SERPINB13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015439653.1 | XP_015295139.1 | 4 - 5 | 798 | 127 | |
| XM_015439654.1 | XP_015295140.1 | 3 - 4 | 1901 | 127 | ||
More Information
Set Membership: |
|

SERPINB13
Genomic Map