Assay Details
| Gene Symbol: SNX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015435250.1 | XP_015290736.1 | 3 - 4 | 1136 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SNX15| Gene Symbol: SNX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015435250.1 | XP_015290736.1 | 3 - 4 | 1136 | 103 | |
Set Membership: |
|