Assay Details
| Gene Symbol: DNMT3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005548558.2 | XP_005548615.1 | 4 - 5 | 652 | 66 | |
| XM_005548559.2 | XP_005548616.1 | 4 - 5 | 654 | 66 | ||
| XM_015446793.1 | XP_015302279.1 | 3 - 4 | 1360 | 66 | ||
| XM_015446794.1 | XP_015302280.1 | 3 - 4 | 969 | 66 | ||
More Information
Set Membership: |
|

DNMT3L
Genomic Map