Assay Details
| Gene Symbol: LOC102144707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005542537.2 | XP_005542594.1 | 6 - 7 | 1531 | 118 | |
| XM_005542538.2 | XP_005542595.1 | 6 - 7 | 951 | 118 | ||
More Information
Set Membership: |
|

LOC102144707
Genomic Map