Assay Details
| Gene Symbol: IMPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005587192.2 | XP_005587249.1 | 6 - 7 | 633 | 100 | |
| XM_005587194.2 | XP_005587251.1 | 6 - 7 | 882 | 100 | ||
| XM_015439880.1 | XP_015295366.1 | 7 - 8 | 716 | 100 | ||
More Information
Set Membership: |
|

IMPA2
Genomic Map